Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, 90% (v/v), Ricca Chemical
SDP

Catalog No. R01890004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01890004A 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R01890004A Supplier Ricca Chemical Company Supplier No. R01890004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-64-1 , 7732-18-5
Molecular Formula C3H6O
Molecular Weight (g/mol) Mixture
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Color Colorless
CAS Min % 86.0, 12.11
CAS Max % 89.0, 12.61
Quantity 4 L
Packaging Natural Poly Bottle
Physical Form Liquid
Boiling Point 56°C
Percent Purity 90%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 90% (v/v) Aqueous
Chemical Name or Material Acetone
Show More Show Less
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.