Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, 99.5% min, ACS, MilliporeSigma™
GSA_VA

Catalog No. MAX01206 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
Poly Drum
Poly-coated Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX01206 500 mL Glass Bottle
MAX01202 1 L Glass Bottle
MAX01209 1 L Poly Bottle
MAX01208 4 L Glass Bottle
MAX0120P4 4 L Poly-coated Glass Bottle
MAX01203 20 L Metal Can
MAX012020 200 L Metal Drum
MAX012030 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MAX01206 Supplier MilliporeSigma Supplier No. US1AX0120/6
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94.2°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 500 mL
Flash Point -18.15°C (-0.67°F) Closed cup
Ignition Point 464.85°C (868.7°F)
Odor Pungent
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max
Assay Percent Range 99.5% min.
Titratable Acid 0.0003meq/g max
Titratable Base 0.0006meq/g max
Linear Formula CH3COCH3
UN Number UN 1090
Water 0.5% max
Boiling Point 56.1°C
Formula Weight 58.08
Percent Purity ≥99.5% (GC, corrected for water content)
Grade ACS/NF/USP
Chemical Name or Material Acetone, For ACS analysis
Show More Show Less
DOTInformation DOT Class 3
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.