Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, 99.6%, ACS reagent

Catalog No. AC423240010
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423240010 1 L EcoSafPak™ Glass Bottle
AC423245000 500 mL EcoSafPak™ Glass Bottle
AC423240025 2.5 L Glass Bottle
AC423240040 4 L EcoSafPak™ Glass Bottle
AC423240250 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC423240010 Supplier Thermo Scientific Chemicals Supplier No. 423240010
  • $277.00
    $98.65 / Each of 1

    Save $178.35

    64% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 67-64-1 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.0°C
CAS Min % 99.5
CAS Max % 100.0
Decomposition Information >4°C
Density 0.7900g/mL
Quantity 1 L
Flash Point -20°C (-4°C)
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3585
Residue after Evaporation 0.001% max.
Specific Gravity 0.79
Assay Percent Range 99.6%
Titratable Acid 0.0003 meq/g max.
Titratable Base 0.0006 meq/g max.
Vapor Pressure Vapor Pressure: 247mbar at 20°C
Linear Formula CH3COCH3
Viscosity 0.32 mPa.s (20°C)
UN Number 1090; Colorless
Water 0.5% max. (K.F.)
Beilstein 01, 635
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Boiling Point 56.0°C
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Formula Weight 58.08
Percent Purity 99.6%
Grade ACS
Chemical Name or Material Acetone
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.