Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, ≥99.9% (GC), Solstice TraceSELECT™

Catalog No. 60047336 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-336 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-336 Supplier Solstice Supplier No. 695081L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TraceSELECT™, for inorganic trace analysis,≥99.9%

  • High purity solvents for speciation analysis by LC-ICP-MS

  • Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance

  • The blank values for metal traces in these solvents are in the ppb range or lower.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Color Colorless
Density 0.791g/mL at 25°C
Quantity 1 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Glass Bottle
Vapor Pressure 184mmHg (20°C)
Linear Formula CH3COCH3
UN Number UN1090
Beilstein 635680
Formula Weight 58.08g/mol
Percent Purity ≥99.9% (GC)
Grade TraceSELECT
Chemical Name or Material Acetone
Show More Show Less
Hazard Statement P210-P280-P304 + P340 + P312-P305 + P351 + P338-P337 + P313-P403 + P235
DOTInformation class 3 - PG 2 - Acetone
RTECSNumber 200-662-2
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.