Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone (ACS AR), Macron Fine Chemicals™

Catalog No. MK244016 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
Packaging:
Glass
Poly
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-244016 4 L Poly bottle
MK-2440-1 1 L Glass
MK-2440-4 4 x 4 L Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MK-244016 Supplier Avantor Supplier No. 244016
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Quantity 4 L
Packaging Poly bottle
Formula Weight 58.08
Grade ACS
Chemical Name or Material Acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.