Learn More
Andwin Scientific Acetone, ACS Grade, 67-64-1, MFCD00008765, 4L Bottle
SDP
Catalog No. NC0208795
Encompass_Preferred
Molecular Formula CH3COCH3, purity Limit ≥99.5%, Molecular Weight, 58.08 g/mol, 4L, Appearance: Clear colorless liquid, Storage Temperature: Ambient
Catalog No. NC0208795 Supplier Andwin Scientific Supplier No. BDH11014LG
Molecular Formula CH3COCH3, purity Limit ≥99.5%, Molecular Weight, 58.08 g/mol, 4L, Appearance: Clear colorless liquid, Storage Temperature: Ambient