Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone ACS Grade, Innovating Science™
SCI_ED

Catalog No. S000490 Shop All Aldon Products
Change view
Click to view available options
Quantity:
354.89 mL
500 mL
1 L
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000490 500 mL
S000494 1 L
S000495 354.89 mL
S000496 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger: Highly flammable liquid and vapor, causes serious eye irritation, may cause drowsiness or dizziness.
Catalog No. S000490 Supplier Aldon Corporation Supplier No. IS10017
Add to Cart
Add to Cart
Danger: Highly flammable liquid and vapor, causes serious eye irritation, may cause drowsiness or dizziness.

Chemical Identifiers

CAS 67-64-1
Molecular Weight (g/mol) 58.08 g/mol

Specifications

Quantity 500 mL
Packaging HDPE
Boiling Point 56°C
Percent Purity 100%
Grade ACS
Chemical Name or Material Acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.