Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone ACS reagent 99.5
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1791244X2.5L
Encompass
Acetone is a polar organic solvent. It can undergo photocatalytic oxidation in the presence of mixed TiO2-rare earth oxides.
Catalog No. 50-289-7547