Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone ACS reagent 99.5
Catalog No. 502903397
Encompass
Acetone is a polar organic solvent. It can undergo photocatalytic oxidation in the presence of mixed TiO2-rare earth oxides.
Catalog No. 50-290-3397 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1791244X4L
Acetone is a polar organic solvent. It can undergo photocatalytic oxidation in the presence of mixed TiO2-rare earth oxides.