Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone ACS reagent 99.5500ML
Catalog No. 502899874
Encompass
Acetone is a polar organic solvent. It can undergo photocatalytic oxidation in the presence of mixed TiO2-rare earth oxides.
Catalog No. 50-289-9874 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1791246X500ML
Acetone is a polar organic solvent. It can undergo photocatalytic oxidation in the presence of mixed TiO2-rare earth oxides.