Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, >99.9% (excluding water), Solstice

Catalog No. 6000219 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-002-19 4 x 4 L Poly-coated Bottle
60-002-18 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-002-19 Supplier Solstice Supplier No. AH0104PC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

CAS Max % 100.0000%
Density 0.79 g/cm3
Quantity 4 x 4 L
Flash Point −20°C
Packaging Poly-coated Bottle
Water 0.5% max.
UN Number UN1090
Beilstein 635680
Boiling Point 56°C
Solubility Information Miscible in Water at 20°C
Formula Weight 58.08
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Acetone, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, >99.9% (excluding water)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.