Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone-Alcohol, 1 + 1 Decolorizer Solution for use in Gram Staining, Reagents

Catalog No. 01335930
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-930 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-930 Supplier Reagents Holdings Llc Supplier No. CA1064004A
Only null left
Add to Cart
Add to Cart

Acetone-Alcohol, 1 + 1 Decolorizer Solution for use in Gram Staining, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Physical Form Liquid
Chemical Name or Material Acetone-Alcohol, 1 + 1 Decolorizer Solution for use in Gram Staining
CAS 67-64-1, 64-17-5, 64-17-5
Boiling Point 60°C
Packaging Poly Bottle
Solubility Completely miscible
Specific Gravity 0.79
Color Colorless
Quantity 4 L
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.