Acetone, AR, For ACS analysis, Macron Fine Chemicals™

macron

Manufacturer: Macron 244010

Quantity 4L

 View more versions of this product

Catalog No. 14-650-254

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 67-64-1
    Physical Form Liquid
    Molecular Formula CH3COCH3
    Formula Weight 58.08
    Chemical Name or Material Acetone, ACS

    Specifications

    Quantity 4L
    Physical Form Liquid
    Color Clear colorless
    Formula Weight 58.08
    Molecular Formula CH3COCH3
    CAS 67-64-1
    Chemical Name or Material Acetone, ACS
    Assay 99.5 % min.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit