Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Capillary GC Grade, ≥99.9%, Solstice

Catalog No. 60046833 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-833 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-833 Supplier Solstice Supplier No. 4146894X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, capillary GC grade,≥99.9%

  • For GC headspace applications
  • Meets USP, Ph. Eur. and ICH requirements
  • GC headspace application tested
  • Packed under inert gas for longer shelf life
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -95°C
CAS 67-64-1
Color Colorless
Density 0.791g/mL at 25°C
Quantity 4 x 4 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3COCH3
Vapor Pressure 184mmHg (20°C)
MDL Number MFCD00008765
UN Number UN1090
Beilstein 635680
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
Solubility Information Completely miscible in water
Boiling Point 55°C to 57°C (1013 hPa)
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08g/mol
Percent Purity ≥99.9%
Chemical Name or Material Acetone
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
DOTInformation class 3 - PG 2 - Acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.