Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone (Certified ACS Grade), Fisher BioReagents

Catalog No. BP24034 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
EcoSafPak™ Amber Glass
Steel Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2403-4 4 L EcoSafPak™ Amber Glass
BP2403-20 20 L Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP2403-4

missing translation for 'validMainframeMsg'

Acetone can be used for biomolecular extraction processes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 7
Melting Point -95°C
Color Colorless
Density 0.790g/cm³
Quantity 4 L
Identification Pass Test
Infrared Spectrum Conforms to reference
Packaging EcoSafPak™ Amber Glass
Physical Form Liquid
Residue after Evaporation 0.001% max.
Titratable Acid 0.0003mEq/g max.
Linear Formula CH3COCH3
Titratable Base 0.0006mEq/g max.
Vapor Pressure 247mbar at 20°C
Viscosity 0.32 mPa/s at 20°C
Water 0.5% max.
Boiling Point 56°C
Formula Weight 58.08
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Acetone
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 3, : Flammable Liquid
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.