Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone CHROMASOLV™, for HPLC, ≥99.8%, Solstice

Catalog No. 6000206 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-002-06 6 x 1 L
60-002-02 1 L
60-002-04 4 x 2.5 L
60-002-05 -
60-002-03 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Repeated exposure may cause skin dryness or cracking.
Catalog No. 60-002-06 Supplier Solstice Supplier No. 348506X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Repeated exposure may cause skin dryness or cracking.

Acetone's luminesence intensity is dependent upon the solution components.1 The absorption of UV light by acetone results in its photolysis and the production of radials.1

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C
Density 0.791g/mL at 25°C
Quantity 6 x 1 L
Refractive Index n20/D 1.359
Vapor Pressure 184mmHg (20°C)
Linear Formula CH3COCH3
Beilstein 635680
Boiling Point 56°C (760 mmHg)
Formula Weight 58.08g/mol
Percent Purity ≥99.8%
Grade CHROMASOLV
Chemical Name or Material Acetone, for HPLC
Show More Show Less
Hazard Statement P210-P280-P304 + P340 + P312-P305 + P351 + P338-P337 + P313-P403 + P235
Precautionary Statement H225-H319-H336
EINECSNumber 200-662-2
RTECSNumber AL3150000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.