Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone-d{6}, 99.9% (Isotopic), contains 1% v/v TMS

Catalog No. AA4245814
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4245814 25 g
AA4245809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4245814 Supplier Thermo Scientific Chemicals Supplier No. 04245814
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 666-52-4 | C3H6O | 64.117 g/mol

Acetone-d{6} is used to produce Perdeuterio-aceton-cyanhydrin at temperature of 15 - 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 666-52-4
Molecular Formula C3H6O
Molecular Weight (g/mol) 64.117
MDL Number MFCD00044635
InChI Key CSCPPACGZOOCGX-WFGJKAKNSA-N
Synonym acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
PubChem CID 522220
ChEBI CHEBI:78217
IUPAC Name 1,1,1,3,3,3-hexadeuteriopropan-2-one
SMILES CC(=O)C

Specifications

Physical Form Liquid
Quantity 25 g
Color Colorless
Density 0.872 g/mL
Odor Acetone-like
Refractive Index 1.3554
Flash Point −17°C (1°F)
Melting Point -94°C
Boiling Point 56°C
UN Number UN1090
Solubility Information Fully miscible in water.
Formula Weight 64.13
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Contains 1% v/v TMS
Chemical Name or Material Acetone-d6, Isotopic
Show More Show Less
Hazard Category H225-H319-H335-H336-H373
Hazard Statement GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONE
EINECSNumber 211-563-9
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.