Learn More
Cambridge Isotope Laboratories Acetone-D6, 666-52-4, MFCD00044635, 25 g
Catalog No. NC9699304
Encompass_Preferred
Chemical Formula: CD3COCD3, MW: 64.12, 99.9%, Synonyms: Propanone; beta-Ketopropane; Dimethylketone.
Catalog No. NC9699304 Supplier Cambridge Isotope Laboratories Supplier No. DLM925
Chemical Formula: CD3COCD3, MW: 64.12, 99.9%, Synonyms: Propanone; beta-Ketopropane; Dimethylketone.