Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone-d6, for NMR, 99.8 atom % D

Catalog No. AC325320500
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325320500 50 mL Glass Bottle
AC325320100 10 mL Glass Bottle
AC325322500 250 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC325320500 Supplier Thermo Scientific Chemicals Supplier No. 325320500
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 666-52-4 | C3H6O | 64.117 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 666-52-4
Molecular Formula C3H6O
Molecular Weight (g/mol) 64.117
MDL Number MFCD00044635
InChI Key CSCPPACGZOOCGX-WFGJKAKNSA-N
Synonym acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
PubChem CID 522220
ChEBI CHEBI:78217
IUPAC Name 1,1,1,3,3,3-hexadeuteriopropan-2-one
SMILES CC(=O)C

Specifications

Physical Form Liquid
Density 0.8700g/mL
Color Colorless
Flash Point −20°C
Quantity 50 mL
Packaging Glass Bottle
Deuterium (D) 99.75% min.
Water 0.1% max. (Coulometric)
Melting Point -93.0°C
Boiling Point 55.0°C
Linear Formula CD3COCD3
Beilstein 01, III, 2741
Formula Weight 64.13
Grade NMR
Chemical Name or Material Acetone-d6, For NMR, 99.8 atom % D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.