Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone, 67-64-1, MFCD00008765, 1 L
Catalog No. NC1140586
Encompass
Linear Formula: CH3COCH3, Molecular Weight: 58.08, for HPLC, ≥99.9%. Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials.
Catalog No. NC1140586 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2707251L
Linear Formula: CH3COCH3, Molecular Weight: 58.08, for HPLC, ≥99.9%. Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials.