Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, For HPLC, 99.9%, MilliporeSigma™ Supelco™

Catalog No. 111005331 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 L
Packaging:
Pure-Pac™ 2 Composite Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-100-5331 50 L Pure-Pac™ 2 Composite Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-5331 Supplier MilliporeSigma Supelco Supplier No. 43912650LP23C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetone is a ketone. Mixtures of TiO2 (P25) with rare earth oxides (La2O3 or Y2O3) calcined at 700°C or 650°C has been reported to efficiently photocatalyze the oxidation of acetone. Its aldol condensation in vapor-phase has been investigated over MgO promoted with alkali (Li, Na, K and Cs) or alkaline earth (Ca, Sr and Ba) metal ions. It undergoes Aldol condensation in the presence of Mg-Al layered double hydroxides (LDH) as catalysts and Cl- and/or CO32- as compensating anions to afford diacetone alcohol and mesityl oxide. Enantioselective Aldol condensation of acetone with various isatins in the presence of dipeptides (catalyst) has been described.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C
Color Colorless
Density 0.791 g/mL (at 25°C)
Quantity 50 L
Packaging Pure-Pac™ 2 Composite Drum
Refractive Index n20/D 1.359
Vapor Pressure 184 mmHg (20°C)
Linear Formula CH3COCH3
Boiling Point 56°C (760 mmHg)
Solubility Information H2O: miscible
Formula Weight 58.08
Percent Purity ≥99.9%
Grade HPLC
Show More Show Less
RTECSNumber AL3150000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.