Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Histological Grade, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18608394 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-394 20 L Steel Can
18-608-386 100 mL Amber Glass Bottle
18-608-392 1 L Amber Glass Bottle
18-608-393 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-608-394 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HS00120LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-64-1

Specifications

CAS Max % 1
Quantity 20 L
Packaging Steel Can
Residue after Evaporation 1e-05
Molecular Formula C3H6O
MDL Number MFCD00008765
Water 0.005
Boiling Point 56°C
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Percent Purity 0.995
Grade Histological
Chemical Name or Material Acetone
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.