Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Solstice

Catalog No. 60090081 Shop All Solstice Research Chemicals Products
Catalog No. 60-090-081 Supplier Solstice Supplier No. CS01156
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (excluding water)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
PubChem CID 180
ChEBI CHEBI:15347

Specifications

Flash Point -20 °C
Packaging Returnable Steel Drum
Physical Form Liquid, Clear
Molecular Formula C3H6O
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.