Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone Laboratory Reagent
Catalog No. 502899163
Encompass
Acetone is a polar organic solvent. It can undergo photocatalytic oxidation in the presence of mixed TiO2-rare earth oxides.
Catalog No. 50-289-9163 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1799734X4L
Acetone is a polar organic solvent. It can undergo photocatalytic oxidation in the presence of mixed TiO2-rare earth oxides.