Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone Laboratory Reagent
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1799734L
Encompass
Acetone is a polar organic solvent. It can undergo photocatalytic oxidation in the presence of mixed TiO2-rare earth oxides.
Catalog No. 50-289-9232