Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Laboratory Reagent, ≥99.5%, Solstice

Catalog No. 60045025 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
4 L
25 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-025 6 x 1 L Glass Bottle
60-045-019 1 L Glass Bottle
60-045-022 4 L Glass Bottle
60-045-023 4 x 4 L Glass Bottle
60-045-021 25 L Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-045-025 Supplier Solstice Supplier No. 1799736X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory Reagent,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C
Color Colorless
Density 0.791g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.359(lit.)
Vapor Pressure 184mmHg (20°C)
Linear Formula CH3COCH3
UN Number UN1090
Beilstein 635680
Boiling Point 56°C
Formula Weight 58.08g/mol
Percent Purity ≥99.5%
Grade Laboratory Reagent
Chemical Name or Material Acetone
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H319-H336
DOTInformation class 3 - PG 2 - Acetone
EINECSNumber 200-662-2
RTECSNumber AL3150000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.