Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone Laboratory Reagent1L
Catalog No. 502898353
Encompass
Acetone is a polar organic solvent. It can undergo photocatalytic oxidation in the presence of mixed TiO2-rare earth oxides.
Catalog No. 50-289-8353 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1799731L
Acetone is a polar organic solvent. It can undergo photocatalytic oxidation in the presence of mixed TiO2-rare earth oxides.