Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, MP Biomedicals™

Catalog No. ICN19383290 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
0.5 L
1 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19383290 0.5 L
ICN19383280 100 mL
MP219383291 1 L
MP219383204 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19383290 Supplier MP Biomedicals, Inc Supplier No. 0219383290
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetone ACS, ≥99.5%, Clear, Colorless Liquid

  • It is a commonly used material in the manufacture of various chemicals like methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid (via the ketene process), iodoform, and bromoform.
  • Other products where acetone is utilized in manufacture include rayon, photographic films, and paint and varnish removers.
  • In chemical synthesis, acetone has been utilized in the synthesis and isolation of both organic and inorganic compounds and complexes.
  • These include t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles, and trispyrazolylborate platinum compounds.
  • Acetone is also used in HPLC analysis of organic and inorganic species, including gold nanoparticle clusters and plant oils. Acetone has been utilized in protein crystallization.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.4°C D396(Literature)
Color ≤10 (APHA)
Density 0.7905 - 0.793g/cm3(Lit.)
Quantity 0.5 L
Flash Point -17°C / 1.4°F
Physical Form Liquid
Refractive Index n20/D 1.359(lit.)
Assay Percent Range ≥99.5%
Titratable Base ≤0.0006 meq/g
Vapor Pressure 184 mmHg (20°C)
Water ≤0.5%
UN Number UN1090
Beilstein 635680EC
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
Boiling Point 56.2°C (at 1013 hPa) (literature)
Solubility Information Soluble in water.
Formula Weight 58.1
Grade ≥99.5%
Chemical Name or Material Acetone, For ACS analysis
Show More Show Less
RTECSNumber AL3150000
storageNote1 Store at Room Temperature(15- 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.