Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, NF, 99%, Spectrum™ Chemical
SDP

Catalog No. 18600278 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-278 200 L Drum
18-600-268 100 mL Amber Glass Bottle
18-600-269 500 mL Amber Glass Bottle
18-600-271 1 L Amber Glass Bottle
18-600-270 6 x 500 mL Amber Glass Bottle
18-600-272 4 L Amber Glass Bottle
18-600-273 4 L Poly Bottle
18-600-274 6 x 1 L Amber Glass Bottle
18-600-275 4 x 4 L Amber Glass Bottle
18-600-276 4 x 4 L Poly Bottle
18-600-277 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18-600-278 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AC115200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AC115, 67-64-1, C3H6O

Spectrum™ Chemical Acetone, NF is a low boiling, volatile organic compound. It is considered one of the five common excipient solvents suitable for pharmaceutical preparations, including water, methanol, dehydrated alcohol and ether. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

CAS Max % 100%
Quantity 200 L
Packaging Drum
Water 0.5%
UN Number UN1090
Percent Purity 99%
Grade NF
Chemical Name or Material Acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.