Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Spectrophotometric Grade, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18608388 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-388 1 L Amber Glass Bottle
18-608-387 500 mL Amber Glass Bottle
18-608-389 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-608-388 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SP1011LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-64-1

Specifications

CAS Max % 1
Quantity 1 L
Packaging Amber Glass Bottle
Residue after Evaporation 1e-05
Titratable Acid 0.0003 meq/g
Titratable Base 0.0006 meq/g
Water 0.005
Boiling Point 56°C
Percent Purity 0.995
Grade Spectrophotometry
Chemical Name or Material Acetone
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.