Learn More
U.S. Pharmacopeia Acetone, 67-64-1, MFCD00008765, 1.5 mL/ampule; 3 ampules
Catalog No. NC9681151
Encompass
Molecular Formula C3H6O, Molecular Weight 58.079, Melting Point -138.46 °F (-94.7 °C), Boiling Point 133.16 °F (56.2 °C), Synonyms: Dimethyl ketone, Ketone propane, 2-Propanone
Catalog No. NC9681151 Supplier U.S. Pharmacopeia Supplier No. 1006801
Molecular Formula C3H6O, Molecular Weight 58.079, Melting Point -138.46 °F (-94.7 °C), Boiling Point 133.16 °F (56.2 °C), Synonyms: Dimethyl ketone, Ketone propane, 2-Propanone