Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, TARR

Catalog No. 502985627
Encompass
Change view
Click to view available options
Quantity:
4 gal
4 x 1 gal.
350 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-298-5627 350 lb.
50-298-5617 4 x 1 gal.
50-298-5598 4 gal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-298-5627 Supplier Thermo Scientific Production Chemicals and Services Supplier No. T10067
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-64-1
Molecular Weight (g/mol) 58.08

Specifications

Quantity 350 lb.
Packaging Drum
Molecular Formula C3H6O
MDL Number MFCD00008765
Boiling Point 132.8 °F
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Grade Semiconductor
Chemical Name or Material Acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.