Learn More
Acetonitrile with 0.1% formic acid, ≥99.5% (GC), Solstice
Shop All Solstice Research Chemicals ProductsSpecifications
Specifications
| Quantity | 4 x 4 L |
| Flash Point | 2°C (35.6°F) |
| UN Number | UN1648 |
| Percent Purity | ≥99.5% (GC) |
| Chemical Name or Material | Acetonitrile with 0.1% formic acid |
Safety and Handling
| Hazard Statement | P210-P261-P302 + P352 + P312-P304 + P340 + P312-P337 + P313-P403 + P235 |
| DOTInformation | 3/PGII |
| RTECSNumber | 200-835-2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.