Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile with 0.1% (v/v) Formic Acid, Hypergrade, For LC-MS LiChrosolv™, MilliporeSigma™

Catalog No. p-7221263
Click to view available options
Quantity:
1 L
2.5 L
4 L

Specifications

Melting Point -45.7°C
Density 0.786 g/cm3 at 20 °C
Boiling Point 81.6°C (1013 hPa)
Quantity 1 L
Explosion Limit 3 to 17% (V)
Flash Point 2 °C
Ignition Point 524 °C
Packaging Glass Bottle
Vapor Pressure 97 hPa at 20 °C
Grade Hypergrade
Recommended Storage Store at 2 to 30 °C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.