Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetonitrile | 75-05-8 | MFCD00001878 | 2.5 L
Catalog No. NC1766039
Encompass
Acetonitrile | Purity: ≥99.9% | Mol Wt: 41.05 | 75-05-8 | MFCD00001878 | 2.5 L
Catalog No. NC1766039 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1000292500
Acetonitrile | Purity: ≥99.9% | Mol Wt: 41.05 | 75-05-8 | MFCD00001878 | 2.5 L