Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Acetonitrile)[(2-Biphenyl)Di-Tert-Butylphosphine]Gold(I) Hexafluoroantimonate

Catalog No. 697575250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-757-5250MG 250 mg
69-757-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-757-5250MG Supplier Sigma Aldrich Supplier No. 697575250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(Acetonitrile)[(2-Biphenyl)Di-Tert-Butylphosphine]Gold(I) Hexafluoroantimonate

Specifications

CAS 866641-66-9
Melting Point 198°C to 203°C
Molecular Formula C22H30AuF6NPSb
Linear Formula C22H30AuF6NPSb
MDL Number MFCD11045449
Quantity 250 mg
Synonym JohnPhos Au(MeCN)SbF6
Molecular Weight (g/mol) 772.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.