Learn More
Electron Microscopy Sciences Acetonitrile 450 ML
SDP
Catalog No. 50980147
Encompass_Preferred
CAS #75-05-8 CH₃CN F.W. 41.05
A non-carcinogenic substitute for Propylene Oxide, used in tissueprocessing for TEM. Good for infiltration with EMbed-812 or other Epon-812 substitutes. Tarnowski, sB.I. et al, EMSA proceedings, Detroit, MI page 38 (1984)
Catalog No. 50-980-147 Supplier Electron Microscopy Sciences Supplier No. 10021
CAS #75-05-8 CH₃CN F.W. 41.05
A non-carcinogenic substitute for Propylene Oxide, used in tissueprocessing for TEM. Good for infiltration with EMbed-812 or other Epon-812 substitutes. Tarnowski, sB.I. et al, EMSA proceedings, Detroit, MI page 38 (1984)