Learn More
GFS Chemicals Inc ACETONITRILE, HPLC, GRADIENT GRADE | 4 X 4 L | GFS CHEMICALS, INC. Item #: 10544 | Bulk #: 2482
SDP
Catalog No. NC1925192
Encompass_Preferred
ACETONITRILE, HPLC, GRADIENT GRADE | Chemical Formula : CH!3CN | CAS # : 75-05-8 | Specifications : Assay 99.9 % - Min, Color (APHA) 10 , LC Gradient Suitability, Residue after evaporation 2 ppm , Titrable acid 0.008 meq/g , Titrable base 0.0006 meq/g , UV Absorbance @ 190 nm 1.00 , UV Absorbance @ 200 nm 0.05 , UV Absorbance @ 210 nm 0.03 , UV Absorbance @ 220 nm 0.02 , UV Absorbance @ 254-400 nm 0.005 , Water 0.01 %
Catalog No. NC1925192 Supplier GFS Chemicals Inc Supplier No. 10544
ACETONITRILE, HPLC, GRADIENT GRADE | Chemical Formula : CH!3CN | CAS # : 75-05-8 | Specifications : Assay 99.9 % - Min, Color (APHA) 10 , LC Gradient Suitability, Residue after evaporation 2 ppm , Titrable acid 0.008 meq/g , Titrable base 0.0006 meq/g , UV Absorbance @ 190 nm 1.00 , UV Absorbance @ 200 nm 0.05 , UV Absorbance @ 210 nm 0.03 , UV Absorbance @ 220 nm 0.02 , UV Absorbance @ 254-400 nm 0.005 , Water 0.01 %