Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile 50%, Water 47.5 % and Trifluoroacetic acid 2.5%, For mass spectrometry, Solstice

Catalog No. 60048220 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-220 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-048-220 Supplier Solstice Supplier No. 19182250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LabReady™ Solvent Blend, for mass spectrometry

LabReady™ Solvent Blends

  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Solstice Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Solstice LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Solstice's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS 76-05-1, 7732-18-5, 75-05-8
Quantity 250 mL
CAS Max % 47.5000%
Flash Point <10°C
Packaging Glass Bottle
UN Number UN1648
Grade Spectrometry
Chemical Name or Material Acetonitrile 50%, Water 47.5% and Trifluoroacetic acid 2.5%, For mass spectrometry
ShelfLife 240 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile solution
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.