Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma Acetonitrile

Catalog No. 01257783 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Acetonitrile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-257-783 Acetonitrile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-783 Supplier MilliporeSigma Supplier No. 1047261000
Add to Cart
Add to Cart

Acetonitrile tested for EPA 533 and EPA 537.1 PFAS methods LiChrosolv™.

  • PFAS tested solvents are used for UHPLC-MS and LC-MS instrumental analysis.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetonitrile
CAS 75-05-8
Quantity 1 L
Packaging Borosilicate Glass Bottle
Boiling Point 81 to 82°C
Molecular Weight (g/mol) 41.05 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.