Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile 80% and Water 20%, Solstice

Catalog No. 60090066 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-066 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-066 Supplier Solstice Supplier No. LC4604X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LabReady™ Solvent Blend, for LC-MS and HPLC

LabReady™ Solvent Blends

  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Solstice Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Solstice LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Solstice's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5, 75-05-8
CAS Max % 24.20%, 75.80%
Quantity 4 x 4 L
Flash Point 8.3°C
Packaging Glass Bottle
Physical Form Liquid, clear
Purity Grade Notes HPLC grade Labready™™ blends
Boiling Point 66.1°C
Molecular Weight (g/mol) 41
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.