Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, BAKER ANALYZED™ ULTRA LC/MS, J.T. Baker™

Catalog No. 02002173 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Flint
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-173 1 L Narrow Mouth Flint
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-173 Supplier Avantor Supplier No. 985302
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For cutting-edge LC/MS applications

    For use in LC/MS UHPLC

Specifications

CAS 75-05-8
Quantity 1 L
Flash Point 2.00°C
Packaging Narrow Mouth Flint
Residue after Evaporation ≤0.8ppm
Specific Gravity 1L = 0.79kg
Molecular Formula C2H3N
Water ≤100ppb
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.05
PubChem CID 6342
ChEBI CHEBI:38472
Formula Weight 41.05
Percent Purity ≥99.9%
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Acetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.