Learn More
VWR INTERNATIONAL INC Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, 75-05-8, MFCD00001878, 4L
Catalog No. NC1641805
Encompass_Preferred
Formula: H₃CCN, ≥99.9%, MW: 41.05 g/mol, Boiling Pt: 81.6 °C (1013 hPa), Melting Pt: –45.7 °C
Catalog No. NC1641805 Supplier VWR INTERNATIONAL INC Supplier No. BDH83639.400
Formula: H₃CCN, ≥99.9%, MW: 41.05 g/mol, Boiling Pt: 81.6 °C (1013 hPa), Melting Pt: –45.7 °C