Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile-d{3}, 99.8% (Isotopic)

Catalog No. AA4226409
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4226409 10 g
AA4226418 50 g
AA4226404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4226409 Supplier Thermo Scientific Chemicals Supplier No. 04226409
Add to Cart

CAS: 2206-26-0 | C2H3N | 44.07 g/mol

Acetonitrile d3 is used as a solvent for NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-26-0
Molecular Formula C2H3N
Molecular Weight (g/mol) 44.07
MDL Number MFCD00001881
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
Synonym acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
PubChem CID 123151
IUPAC Name 2,2,2-trideuterioacetonitrile
SMILES [2H]C([2H])([2H])C#N

Specifications

Physical Form Liquid
Color Colorless
Quantity 10 g
Density 0.844 g/mL
Flash Point 5°C (42°F)
Odor Pungent
Sensitivity Hygroscopic
Boiling Point 81°C
UN Number UN1648
Merck Index 14,70
Solubility Information Miscible with water,ethanol,diethyl ether,acetone,benzene,methanol,methyl acetate,ethyl acetate,acetamide solutions,chloroform,carbon tetrachloride,ethylene chloride and unsaturated hydrocarbons. Immiscible with saturated hydrocarbons.
Formula Weight 44.08
Percent Purity 99.8%
Chemical Name or Material Acetonitrile-d3, Isotopic
Show More Show Less
Hazard Category H225-H302+H312+H332-H319
Hazard Statement GHS H Statement
H225-H302-H312-H332-H319
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE
EINECSNumber 218-616-5
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.