Learn More
Cambridge Isotope Laboratories Acetonitrile-D3, 2206-26-0, Mfcd00001881, 10X0.75 Ml
Catalog No. NC9931208
Encompass_Preferred
Chemical Formula: CD3CN, MW: 44.07, 99.8%, Synonyms: Methyl cyanide; Cyanomethane; Ethanenitrile.
Catalog No. NC9931208 Supplier Cambridge Isotope Laboratories Supplier No. DLM2110X0.75
Chemical Formula: CD3CN, MW: 44.07, 99.8%, Synonyms: Methyl cyanide; Cyanomethane; Ethanenitrile.