Learn More
Cambridge Isotope Laboratories Acetonitrile-D3, 2206-26-0, MFCD00001881, 25 g
Catalog No. NC9687122
Encompass_Preferred
Chemical Formula: CD3CN, MW: 44.07, 99.8%, Synonyms: Methyl cyanide; Cyanomethane; Ethanenitrile.
Catalog No. NC9687122 Supplier Cambridge Isotope Laboratories Supplier No. DLM2125
Chemical Formula: CD3CN, MW: 44.07, 99.8%, Synonyms: Methyl cyanide; Cyanomethane; Ethanenitrile.