Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile - D3, deuteration degree min. 99% for NMR spectroscopy MagniSolv™, MilliporeSigma™

Catalog No. M1029040010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Septum Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1029040010 10 mL Septum Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1029040010 Supplier MilliporeSigma Supplier No. 1.02904.0010
Add to Cart
Add to Cart

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-26-0
Molecular Formula C2H3N
Molecular Weight (g/mol) 44.07
MDL Number MFCD00001881
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
Synonym acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
PubChem CID 123151
IUPAC Name (²H₃)acetonitrile
SMILES [2H]C([2H])([2H])C#N

Specifications

Physical Form Liquid
Odor Ether-like
Quantity 10 mL
Viscosity 0.39mPa.s (20°C)
Packaging Septum Ampule
Vapor Pressure 97 hPa at 20°C (68°F)
Assay Percent Range 99% min.
Formula Weight 44.07
Chemical Name or Material Acetonitrile - D3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.