Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile-d3, for NMR, 99.8 atom % D

Catalog No. AC464160100
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464160100 10 mL
AC464160250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC464160100 Supplier Thermo Scientific Chemicals Supplier No. 464160100
Add to Cart
This item is not returnable. View return policy

CAS: 2206-26-0 | C2H3N | 44.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-26-0
Molecular Weight (g/mol) 44.07

Specifications

Refractive Index 1.3430 to 1.3450
Density 0.8440 g/mL
Viscosity 0.39 mPa.s (20°C)
Specific Gravity 0.844
Flash Point 2°C
Quantity 10 mL
Melting Point -46.0°C
Boiling Point 79.0°C to 81.0°C
Molecular Formula C2D3N
Linear Formula CD3CN
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 44.07
Grade NMR
Chemical Name or Material Acetonitrile-d3
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Highly flammable liquid and vapor.

Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Causes serious eye irritation.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF ON SKIN: Wash with plenty of soap and water.

IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

EINECSNumber 218-616-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.