Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile-d3, for NMR, 99 atom % D

Catalog No. AC166230250
Change view
Click to view available options
Quantity:
10 mL
25 mL
50 mL
250 mL
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166230250 25 mL Glass Bottle
AC166230100 10 mL Glass Bottle
AC166230500 50 mL Glass Bottle
AC166232500 250 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC166230250 Supplier Thermo Scientific Chemicals Supplier No. 166230250
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acetonitrile-d3, C2H3N, CAS Number-2206-26-0, 2h3 acetonitrile, acetonitrile-2,2,2-d3, acetonitrile-d3, acetonitrile-d3, >-99.8 atom % d, acetonitrile-d3, 99.8 atom % d, acetonitrile-d isotopic 5g, cd3cn, methyl-d3 cyanide, trideuteroacetonitrile, 10mL, 02, IV, 428, 79.0 deg.C to 81.0 deg.C | CAS: 2206-26-0 | C2H3N | 44.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-26-0
Molecular Formula C2H3N
Molecular Weight (g/mol) 44.07
MDL Number MFCD00001881
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
PubChem CID 123151
SMILES [2H]C([2H])([2H])C#N

Specifications

Physical Form Liquid
Color Colorless
Viscosity 0.39 mPa.s (20°C)
Packaging Glass Bottle
Density 0.8440g/mL
Flash Point 2°C
Refractive Index 1.3430 to 1.3450
Quantity 25 mL
Water 0.1% max. (Coulometric)
Deuterium (D) 98.5% min.
Melting Point -46.0°C
Boiling Point 79.0°C to 81.0°C
Linear Formula CD3CN
Beilstein 02, IV, 428
Synonym acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
IUPAC Name 2,2,2-trideuterioacetonitrile
Formula Weight 44.07
Grade NMR
Chemical Name or Material Acetonitrile-d3, 99 atom%D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.