Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile-d3, Thermo Scientific™

Catalog No. AC214520010
Change view
Click to view available options
Quantity:
10 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC214520010 10 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC214520010 Supplier Thermo Scientific Chemicals Supplier No. 214520010

missing translation for 'validMainframeMsg'

For NMR

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-26-0
Molecular Formula C2H3N
Molecular Weight (g/mol) 44.07
MDL Number MFCD00001881
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
Synonym acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, >=99.8 atom % d, acetonitrile-d isotopic 5g
PubChem CID 123151
IUPAC Name (²H₃)acetonitrile
SMILES [2H]C([2H])([2H])C#N

Specifications

Melting Point -46.0°C
Density 0.8440g/mL
Boiling Point 79.0°C to 81.0°C
Flash Point 2°C
Refractive Index 1.3430 to 1.3450
Linear Formula CD3CN
Quantity 10 x 0.5 mL
Specific Gravity 0.844
Solubility Information Solubility in water: miscible.
Formula Weight 44.07
Grade NMR
Chemical Name or Material Acetonitrile-d3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.